missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRM1 Mouse anti-Human, Mouse, Rat, Clone: 2D3B6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66763-1-IG-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene mediates leucine-rich nuclear export signal (NES)-dependent protein transport. Exportin 1 specifically inhibits the nuclear export of Rev and U snRNAs. It is involved in the control of several cellular processes by controlling the localization of cyclin B, MPAK, and MAPKAP kinase 2. This protein also regulates NFAT and AP-1.Specifications
| CRM1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O14980, Q6P5F9, Q80U96 | |
| Xpo1 | |
| CRM1 Fusion Protein Ag27337 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 2D3B6 | |
| Unconjugated | |
| XPO1 | |
| CRM1, DKFZp686B1823, Exp1, Exportin 1, XPO1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 103573, 7514, 85252 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction