missing translation for 'onlineSavingsMsg'
Learn More

CRM1 Mouse anti-Human, Mouse, Rat, Clone: 2D3B6, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen CRM1
Clone 2D3B6
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16888935
View Documents
Proteintech
66763-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16878935
View Documents
Proteintech
66763-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The protein encoded by this gene mediates leucine-rich nuclear export signal (NES)-dependent protein transport. Exportin 1 specifically inhibits the nuclear export of Rev and U snRNAs. It is involved in the control of several cellular processes by controlling the localization of cyclin B, MPAK, and MAPKAP kinase 2. This protein also regulates NFAT and AP-1.
Specifications

Specifications

CRM1
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
CRM1, DKFZp686B1823, Exp1, Exportin 1, XPO1
Xpo1
IgG1
Protein G
Antibody
2D3B6
Western Blot
Unconjugated
Mouse
Rat, Mouse, Human
O14980, Q6P5F9, Q80U96
103573, 7514, 85252
CRM1 Fusion Protein Ag27337
Primary
-20°C
XPO1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.