missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRM1 Mouse, anti-Human, CoraLite Plus 488, Clone: 2D3B6, Proteintech
Mouse Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | CRM1 |
|---|---|
| Clone | 2D3B6 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18027295
|
Proteintech
CL488-66763-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18030336
|
Proteintech
CL488-66763-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CRM1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CRM1. This antibody recognizes human, mouse, rat antigen. The CRM1 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
| CRM1 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 7514 | |
| IgG1 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 2D3B6 | |
| Western Blot | |
| Liquid | |
| Human, Mouse, Rat | |
| CRM1, DKFZp686B1823, Exp1, Exportin 1, XPO1 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title