Learn More
CRM1 Mouse, anti-Human, CoraLite Plus 488, Clone: 2D3B6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66763-20UL
Description
The CRM1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CRM1. This antibody recognizes human, mouse, rat antigen. The CRM1 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
| CRM1 | |
| 2D3B6 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| 155 kDa | |
| IgG1 |
| Western Blot | |
| 1000 μg/mL | |
| CRM1, DKFZp686B1823, Exp1, Exportin 1, XPO1 | |
| 20 μL | |
| 7514 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |