missing translation for 'onlineSavingsMsg'
Learn More

CRISPR-Cas9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£280.00 - £401.00

Specifications

Antigen CRISPR-Cas9
Concentration 1 mg/mL
Dilution Western Blot 1:1000-1:10000, Immunocytochemistry/ Immunofluorescence :1000-1:2000
Applications Western Blot, Immunofluorescence
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18636596
View Documents
Novus Biologicals
NBP3-05548-100ul
100 μg
£401.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18656877
View Documents
Novus Biologicals
NBP3-05548-50ul
50 μg
£280.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CRISPR-Cas9 Polyclonal antibody specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

CRISPR-Cas9
Western Blot 1:1000-1:10000, Immunocytochemistry/ Immunofluorescence :1000-1:2000
Polyclonal
Purified
RUO
Bacteria
Cas9, CRISPR, CRISPR-associated protein 9 nuclease
N-terminal region, amino acids 1-608 and C-terminal region, amino acids 814-1372 of S. pyogenes CRISPR-CAS9 sequence CDJ55032.1 from S. pyogenes, expressed in and purified from E. coli
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
1 mg/mL
Western Blot, Immunofluorescence
Unconjugated
Rabbit
Gene Editing, Gene Knockouts, Gene Therapy
50% PBS, 50% glycerol
901176
IgG
Immunogen affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.