missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRISPR-Cas9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05548-50ul
This item is not returnable.
View return policy
Description
CRISPR-Cas9 Polyclonal antibody specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| CRISPR-Cas9 | |
| Polyclonal | |
| Unconjugated | |
| 50% PBS, 50% glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Bacteria | |
| Purified |
| Western Blot, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1:1000-1:10000, Immunocytochemistry/ Immunofluorescence :1000-1:2000 | |
| Cas9, CRISPR, CRISPR-associated protein 9 nuclease | |
| N-terminal region, amino acids 1-608 and C-terminal region, amino acids 814-1372 of S. pyogenes CRISPR-CAS9 sequence CDJ55032.1 from S. pyogenes, expressed in and purified from E. coli | |
| 50 μg | |
| Gene Editing, Gene Knockouts, Gene Therapy | |
| 901176 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction