missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRIPT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11211-1-AP-20UL
This item is not returnable.
View return policy
Description
Involved in the cytoskeletal anchoring of DLG4 in excitatory synapses.Specifications
| CRIPT | |
| Polyclonal | |
| Unconjugated | |
| CRIPT | |
| CRIPT, HSPC139 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56724, 56725, 9419 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O70333, Q792Q4, Q9P021 | |
| Cript | |
| CRIPT Fusion Protein Ag1708 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction