missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRIPT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CRIPT |
|---|---|
| Concentration | 0.47 mg/mL |
| Applications | Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16808143
|
Proteintech
11211-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16898133
|
Proteintech
11211-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Involved in the cytoskeletal anchoring of DLG4 in excitatory synapses.Specifications
| CRIPT | |
| Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| O70333, Q792Q4, Q9P021 | |
| 56724, 56725, 9419 | |
| CRIPT Fusion Protein Ag1708 | |
| Primary | |
| -20°C | |
| CRIPT |
| 0.47 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| CRIPT, HSPC139 | |
| Cript | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title