Learn More
Fisherbrand™ Crimping Heads for Pneumatic Hand-held Crimping Tool
Product Code. 12980981 Shop All Fisherbrand Products
Click to view available options
Description:
11mm Crimping Head
13mm Crimping Head
20mm Crimping Head
32mm Crimping Head
7.5mm Crimping Head
8mm Crimping Head
For Use With (Equipment):
Aluminum Cap
Aluminum/Steel Caps
Flip Top/Flip Off Seals
Unit Size:
Each
Product Code. 12980981
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use with Pneumatic Hand-held Crimping Tool.
  • Crimping Head
  • Pneumatic
  • Steel
TRUSTED_SUSTAINABILITY
Specifications
Description 20mm Crimping Head
For Use With (Equipment) Flip Top/Flip Off Seals