Learn More
DWK Life Sciences Crimp Cap, ND11
Product Code. 16606872 Shop All DWK Life Sciences Products
Change view
Click to view available options
Material:
Natural Red Rubber, Clear FEP
Red PTFE, White Silicone
Red Rubber, Clear PTFE
Red Rubber, White PTFE
White Silicone, Red PTFE
Unit Size:
Pack of 100
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
16606872 Natural Red Rubber, Clear FEP Pack of 100
16686862 White Silicone, Red PTFE Pack of 100
16696862 Red PTFE, White Silicone Pack of 100
16616872 Red Rubber, Clear PTFE Pack of 100
16626872 Red Rubber, White PTFE Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16606872 Supplier DWK Life Sciences Supplier No. 225683
Only null left
Add to basket
Edge
Add to basket
Crimp closures generally known for excellent sealing characteristics, even when it comes to long-term sample storage.
The quality of a crimp seal is strongly dependent on the right calibration of the crimping tool used to apply the closure to the vial. A too tight or too loose crimp will impair the quality of the seal. To maintain good results, make sure to re-calibrate your crimping tool regularly and check the quality of the seal every time you crimp.
  • Packed in zip-lock PE-bags with tear-off foil strip to ensure tamper-evidence and re-closeability.
TRUSTED_SUSTAINABILITY
Specifications
Color Silver
Material Natural Red Rubber, Clear FEP
Includes With liner
Closure Type Open Top Cap
Closure Size ND20