missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRHR2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25267-1-AP-20UL
This item is not returnable.
View return policy
Description
Corticotropin-releasing hormone (CRH; 122560) is a 41-amino acid peptide synthesized in the hypothalamus, and play a role in the neuroregulation, responses to stress and immune challenge. Corticotropin Releasing Factor Receptor 2 (CRHR2) was the receptor of CRH. The suppression of CRHR2 has been reported in various regions of the brain as well as in placenta, umbilical vein, heart, epididymis, gastrointestinal tract, adrenal and skeletal muscle.Specifications
| CRHR2 | |
| Polyclonal | |
| Unconjugated | |
| CRHR2 | |
| CRF R 2, CRF R2, CRF2R, CRFR 2, CRFR2, CRH R 2, CRH R2, CRH2R, CRHR2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12922, 1395, 64680 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P47866, Q13324, Q60748 | |
| Crhr2 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering