missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRHR2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25267-1-AP-150UL
This item is not returnable.
View return policy
Description
Corticotropin-releasing hormone (CRH; 122560) is a 41-amino acid peptide synthesized in the hypothalamus, and play a role in the neuroregulation, responses to stress and immune challenge. Corticotropin Releasing Factor Receptor 2 (CRHR2) was the receptor of CRH. The suppression of CRHR2 has been reported in various regions of the brain as well as in placenta, umbilical vein, heart, epididymis, gastrointestinal tract, adrenal and skeletal muscle.Specifications
| CRHR2 | |
| Polyclonal | |
| Unconjugated | |
| CRHR2 | |
| CRF R 2, CRF R2, CRF2R, CRFR 2, CRFR2, CRH R 2, CRH R2, CRH2R, CRHR2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12922, 1395, 64680 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P47866, Q13324, Q60748 | |
| Crhr2 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction