missing translation for 'onlineSavingsMsg'
Learn More

CRHR1/CRF1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£159.00 - £359.00

Specifications

Antigen CRHR1/CRF1
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18630508
View Documents
Novus Biologicals
NBP3-05630-100ul
100 μg
£359.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18671177
View Documents
Novus Biologicals
NBP3-05630-20ul
20 μg
£159.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CRHR1/CRF1 Polyclonal antibody specifically detects CRHR1/CRF1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

CRHR1/CRF1
Western Blot
Unconjugated
Rabbit
GPCR, Neuroscience, Neurotransmission
PBS with 50% glycerol, pH7.3.
1394
IgG
Affinity purified
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
Human, Mouse, Rat
corticotropin releasing hormone receptor 1, corticotropin-releasing factor receptor 1, corticotropin-releasing factor type 1 receptor, Corticotropin-releasing hormone receptor 1, CRF1, CRFR, CRF-R, CRFR1, CRF-R1, CRF-R-1, CRFR-1, CRH-R-1, CRHR1f, CRH-R1h, CRHR1L, CRHRCRH-R1, seven transmembrane helix receptor
Recombinant fusion protein containing a sequence corresponding to amino acids 24-120 of human CRHR1/CRF1 (NP_001138618.1). SLQDQHCESLSLASNISGLQCNASVDLIGTCWPRSPAGQLVVRPCPAFFYGVRYNTTNNGYRECLANGSWAARVNYSECQEILNEEKKSKVHYHVAV
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.