missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRELD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29903-1-AP-20UL
This item is not returnable.
View return policy
Description
CRELD1 Polyclonal antibody specifically detects CRELD1 in Human, Mouse, Rat samples. It is validated for IF, IHC, WB, ELISASpecifications
| CRELD1 | |
| Polyclonal | |
| Unconjugated | |
| NM_080725 | |
| Antigen affinity purification | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunofluorescence, Immunohistochemistry, Western Blot, ELISA | |
| 1400 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| Rabbit | |
| 20 μL | |
| 78987 | |
| Store at -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction