missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRELD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CRELD1 |
|---|---|
| Concentration | 1400 μg/mL |
| Applications | Immunofluorescence, Immunohistochemistry, Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17701129
|
Proteintech
29903-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17791119
|
Proteintech
29903-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CRELD1 Polyclonal antibody specifically detects CRELD1 in Human, Mouse, Rat samples. It is validated for IF, IHC, WB, ELISASpecifications
| CRELD1 | |
| Immunofluorescence, Immunohistochemistry, Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| IgG | |
| Antigen affinity purification | |
| Antibody |
| 1400 μg/mL | |
| Polyclonal | |
| Liquid | |
| Human, Mouse, Rat | |
| 78987 | |
| Primary | |
| Store at -20°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title