missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CREB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22516-0.025mg
This item is not returnable.
View return policy
Description
CREB Polyclonal specifically detects CREB in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CREB | |
| Polyclonal | |
| Unconjugated | |
| PBS with 1 mg/mL BSA with 0.05% Sodium Azide | |
| CREB1 | |
| Synthetic peptide corresponding to residues K(123) R R E I L S R R P S Y R K(136) of human CREB. | |
| 0.025 mg | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 2 μg/mL, Immunohistochemistry 1:20 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunohistochemistry-Paraffin 1:20 - 1:200, Chromatin Immunoprecipitation (ChIP) 1-3 ul | |
| active transcription factor CREB, cAMP responsive element binding protein 1, cAMP-response element-binding protein-1, cAMP-responsive element-binding protein 1, CREB, CREB-1, cyclic AMP-responsive element-binding protein 1, MGC9284, transactivator protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 1385 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction