missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CREB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£181.00
Specifications
| Antigen | CREB |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2 μg/mL, Immunohistochemistry 1:20 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunohistochemistry-Paraffin 1:20 - 1:200, Chromatin Immunoprecipitation (ChIP) 1-3 ul |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
Description
CREB Polyclonal specifically detects CREB in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| CREB | |
| Western Blot 2 μg/mL, Immunohistochemistry 1:20 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunohistochemistry-Paraffin 1:20 - 1:200, Chromatin Immunoprecipitation (ChIP) 1-3 ul | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| active transcription factor CREB, cAMP responsive element binding protein 1, cAMP-response element-binding protein-1, cAMP-responsive element-binding protein 1, CREB, CREB-1, cyclic AMP-responsive element-binding protein 1, MGC9284, transactivator protein | |
| CREB1 | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS with 1 mg/mL BSA with 0.05% Sodium Azide | |
| 1385 | |
| Synthetic peptide corresponding to residues K(123) R R E I L S R R P S Y R K(136) of human CREB. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title