missing translation for 'onlineSavingsMsg'
Learn More

Creatine Phosphate Disodium Salt Tetrahydrate, ≥98%, Thermo Scientific™

Product Code. 15815118 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Plastic bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15815118 5 g Plastic bottle 5g
15825118 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15815118 Supplier Thermo Scientific Supplier No. J13920.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71519-72-7
Molecular Formula C4H8N3Na2O5P
Molecular Weight (g/mol) 255.08
MDL Number MFCD00150192
InChI Key RNTXMYSPASRLFT-UHFFFAOYSA-L
Synonym creatine phosphate sodium salt, Sodium creatine phosphate dibasic tetrahydrate
PubChem CID 131887655
IUPAC Name disodium tetrahydrate ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate
SMILES [Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 5 g
Formula Weight 327.14
Percent Purity ≥98%
Packaging Plastic bottle
Chemical Name or Material Creatine Phosphate Disodium Salt Tetrahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.