accessibility menu, dialog, popup

UserName

H3CNOHONPOOONH2H2OH2OH2OH2ONaNa(E/Z)

CAS RN 71519-72-7

IUPAC Name: disodium tetrahydrate ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate
Synonyms: disodium tetrahydrate {amino[(carboxymethyl)(methyl)amino]methylidene}aminophosphonate
Molecular Weight (g/mol): 327.14
Molecular Formula: C4H16N3Na2O9P
InChi Key: HUWYWJSJJDCZRQ-UHFFFAOYSA-L
SMILES: O.O.O.O.[Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O
Percent Purity 
  • (5)
  • (3)
Physical Form 
  • (2)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (5)
  • (3)
  • (2)
  • (6)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)