Learn More
Creatine phosphate disodium salt tetrahydrate, 98+%
Description
Creatine phosphate disodium salt acts as a substrate for creatine kinase determinations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Creatine phosphate disodium salt acts as a substrate for creatine kinase determinations.
Solubility
Soluble in water.0.1 g/mL.
Notes
Incompatible with Strong bases. strong acids. Protect from Direct sunlight.
Specifications
Specifications
| Assay Percent Range | ≥98% |
| Quantity | 1 g |
| Solubility Information | Soluble in water.0.1g/mL. |
| Formula Weight | 327.14 (255.07 Anhydrous) |
| Percent Purity | ≥98% |
| Physical Form | Powder |
| Chemical Name or Material | Creatine Phosphate Disodium Salt Tetrahydrate |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.