missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine Kinase, Muscle/CKMM Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-52864
This item is not returnable.
View return policy
Description
Creatine Kinase, Muscle/CKMM Polyclonal specifically detects Creatine Kinase, Muscle/CKMM in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Creatine Kinase, Muscle/CKMM | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CKMMcreatine kinase M-type, Creatine Kinase Isoenzyme, Creatine kinase M chain, creatine kinase, muscle, creatine kinase-M, EC 2.7.3, EC 2.7.3.2, M-CK | |
| Rabbit | |
| 43 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P06732 | |
| CKM | |
| Synthetic peptides corresponding to CKM(creatine kinase, muscle) The peptide sequence was selected from the middle region of CKM. Peptide sequence GVDTAAVGSVFDVSNADRLGSSEVEQVQLVVDGVKLMVEMEKKLEKGQSI. | |
| Affinity purified | |
| RUO | |
| 1158 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction