missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPXCR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80386
This item is not returnable.
View return policy
Description
CPXCR1 Polyclonal specifically detects CPXCR1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CPXCR1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CPX chromosomal region candidate gene 1 protein, CPX chromosome region, candidate 1, CT77Cancer/testis antigen 77 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 53336 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| NP_149037 | |
| CPXCR1 | |
| Synthetic peptide directed towards the middle region of human CPXCR1. Peptide sequence WANQLAAVAAGARVAGTQACATETIDTSRVSLRAPQEFMTSHSEAGSRIV. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Crab-eating macaque: 85%. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction