missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPT1C Mouse anti-Human, Porcine, Clone: 5D11B5, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CPT1C |
|---|---|
| Clone | 5D11B5 |
| Concentration | 0.98 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16857615
|
Proteintech
66072-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16847615
|
Proteintech
66072-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody can bind the close sequences genes.
This gene encodes a member of the carnitine/choline acetyltransferase family. The encoded protein regulates the beta-oxidation and transport of long-chain fatty acids into mitochondria, and may play a role in the regulation of feeding behavior and whole-body energy homeostasis. Alternatively spliced transcript variants encoding multiple protein isoforms have been observed for this gene.Specifications
| CPT1C | |
| 0.98 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CATL1, CPT IC, CPT1 B, CPT1C, CPT1P, CPTI B, CPTIC | |
| CPT1C | |
| IgG2b | |
| Protein A | |
| Antibody |
| 5D11B5 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Pig | |
| Q8TCG5 | |
| 100519258, 126129 | |
| CPT1C Fusion Protein Ag17963 | |
| Primary | |
| -20°C | |
| CPT1C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title