missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPT1C Mouse anti-Human, Porcine, Clone: 5D11B5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66072-1-IG-150UL
This item is not returnable.
View return policy
Description
This antibody can bind the close sequences genes.
This gene encodes a member of the carnitine/choline acetyltransferase family. The encoded protein regulates the beta-oxidation and transport of long-chain fatty acids into mitochondria, and may play a role in the regulation of feeding behavior and whole-body energy homeostasis. Alternatively spliced transcript variants encoding multiple protein isoforms have been observed for this gene.Specifications
| CPT1C | |
| Monoclonal | |
| 0.98 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8TCG5 | |
| CPT1C | |
| CPT1C Fusion Protein Ag17963 | |
| 150 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| 5D11B5 | |
| Unconjugated | |
| CPT1C | |
| CATL1, CPT IC, CPT1 B, CPT1C, CPT1P, CPTI B, CPTIC | |
| Mouse | |
| Protein A | |
| RUO | |
| 100519258, 126129 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction