missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPT1C Mouse anti-Human, Porcine, Clone: 5D11B5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66072-1-IG-20UL
This item is not returnable.
View return policy
Description
This antibody can bind the close sequences genes.
This gene encodes a member of the carnitine/choline acetyltransferase family. The encoded protein regulates the beta-oxidation and transport of long-chain fatty acids into mitochondria, and may play a role in the regulation of feeding behavior and whole-body energy homeostasis. Alternatively spliced transcript variants encoding multiple protein isoforms have been observed for this gene.Specifications
| CPT1C | |
| Monoclonal | |
| 0.98 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8TCG5 | |
| CPT1C | |
| CPT1C Fusion Protein Ag17963 | |
| 20 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| 5D11B5 | |
| Unconjugated | |
| CPT1C | |
| CATL1, CPT IC, CPT1 B, CPT1C, CPT1P, CPTI B, CPTIC | |
| Mouse | |
| Protein A | |
| RUO | |
| 100519258, 126129 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido