missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPSF73 Antibody (SR2209), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | CPSF73 |
|---|---|
| Clone | SR2209 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18671434
|
Novus Biologicals
NBP3-22470-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18613704
|
Novus Biologicals
NBP3-22470-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CPSF73 Monoclonal antibody specifically detects CPSF73 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
| CPSF73 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 51692 | |
| IgG | |
| Affinity purified |
| SR2209 | |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| cleavage and polyadenylation specific factor 3, 73kD subunit, cleavage and polyadenylation specific factor 3, 73kDa, cleavage and polyadenylation specificity factor 3, Cleavage and polyadenylation specificity factor 73 kDa subunit, cleavage and polyadenylation specificity factor subunit 3, CPSF 73 kDa subunit, CPSF-73, CPSF73CPSF, EC 3.1.27, EC 3.1.27.-, mRNA 3'-end-processing endonuclease CPSF-73, YSH1 | |
| A synthesized peptide derived from human CPSF73 (Uniprot #: Q9UKF6) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title