missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CPM (7-Diethylamino-3-(4'-Maleimidylphenyl)-4-Methylcoumarin)

Product Code. 11590296
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11590296 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11590296 Supplier Invitrogen™ Supplier No. D346
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

The thiol-reactive coumarin, CPM is very weakly fluorescent until reacted with thiols producing a conjugate with excitation/emission maxima of ∼384/470 nm.

The thiol-reactive coumarin, CPM is very weakly fluorescent until reacted with thiols producing a conjugate with excitation/emission maxima of ∼384/470 nm.

TRUSTED_SUSTAINABILITY

Specifications

Description CPM (7-Diethylamino-3-(4'-Maleimidylphenyl)-4-Methylcoumarin), 25mg, Solid, Methylcoumarin dye, 402.45 Molecular weight, Thiol Chemical Reactivity, 384/470nm Excitation/Emission Wavelength, Used for Labeling Chemistry, Protein Labeling and Crosslinking
Formulation Solid
For Use With (Application) Labeling chemistry, protein labeling and crosslinking
Quantity 25 mg
Product Type CPM
Molecular Weight (g/mol) 402.45kDa
Content And Storage Store in freezer (-5°C to -30°C) and protect from light.
Chemical Reactivity Thiol
Shipping Condition Room Temperature
Reactive Moiety Maleimide
Label or Dye Methylcoumarin
Label Type Classic Dyes
Color Yellow
Emission 470 nm
Excitation 384 nm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.