missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPI17 alpha Antibody (SR1255), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | CPI17 alpha |
|---|---|
| Clone | SR1255 |
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:30 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625844
|
Novus Biologicals
NBP3-21616-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18613104
|
Novus Biologicals
NBP3-21616-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CPI17 alpha Monoclonal antibody specifically detects CPI17 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| CPI17 alpha | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:30 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 17 kDa PKC-potentiated inhibitory protein of PP1, CPI-1717-kDa PKC-potentiated inhibitory protein of PP1, CPI1717-KDa protein, PKC-potentiated inhibitory protein of PP1, PPP1INL, Protein kinase C-potentiated inhibitor protein of 17 kDa, protein phosphatase 1 regulatory subunit 14A, protein phosphatase 1, regulatory (inhibitor) subunit 14A | |
| A synthesized peptide derived from human CPI17 alpha (Uniprot #: Q96A00) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1255 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific, Protein Phosphatase | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 94274 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title