missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPI17 alpha Antibody (SR1255), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21616-100ul
This item is not returnable.
View return policy
Description
CPI17 alpha Monoclonal antibody specifically detects CPI17 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| CPI17 alpha | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SR1255 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:30 | |
| 17 kDa PKC-potentiated inhibitory protein of PP1, CPI-1717-kDa PKC-potentiated inhibitory protein of PP1, CPI1717-KDa protein, PKC-potentiated inhibitory protein of PP1, PPP1INL, Protein kinase C-potentiated inhibitor protein of 17 kDa, protein phosphatase 1 regulatory subunit 14A, protein phosphatase 1, regulatory (inhibitor) subunit 14A | |
| A synthesized peptide derived from human CPI17 alpha (Uniprot #: Q96A00) | |
| 100 μg | |
| Phospho Specific, Protein Phosphatase | |
| 94274 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction