missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 29 publications
£207.00 - £594.00
Specifications
| Antigen | COX-2 |
|---|---|
| Dilution | Western Blot 1:100-1:2000, Simple Western 1:100, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18776193
|
Novus Biologicals
NB100-689SS |
0.1 mL |
£207.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18789663
|
Novus Biologicals
NB100-689 |
£594.00
0.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
COX-2 Polyclonal specifically detects COX-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| COX-2 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| RUO | |
| Human, Rat | |
| P35355 | |
| 5743 | |
| Recombinant protein corresponding to the C-terminus of rat COX2 (the exact amino acid sequence is considered to be proprietary information). | |
| Primary | |
| Store at 4C. Do not freeze. |
| Western Blot 1:100-1:2000, Simple Western 1:100, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cell Biology, Cellular Markers, Cytokine Research, Hypoxia, Lipid and Metabolism, Signal Transduction | |
| PBS (pH 7.4), 0.2% BSA, Tween-20 with 0.05% Sodium Azide | |
| COX-2, COX2cyclooxygenase 2b, cyclooxygenase-2, EC 1.14.99, EC 1.14.99.1, GRIPGHS, hCox-2, PGG/HS, PGH synthase 2, PGHS-2, PHS II, PHS-2, prostaglandin G/H synthase 2, prostaglandin G/H synthase and cyclooxygenase, Prostaglandin H2 synthase 2, Prostaglandin-endoperoxide synthase 2, prostaglandin-endoperoxide synthase 2 (prostaglandin G/H synthase andcyclooxygenase) | |
| PTGS2 | |
| IgG | |
| Affinity Purified | |
| This reacts with the 70 kDa protein. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title