missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 29 publications
Brand: Novus Biologicals NB100-689SS
This item is not returnable.
View return policy
Description
COX-2 Polyclonal antibody specifically detects COX-2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications
| COX-2 | |
| Polyclonal | |
| Western Blot 1:100-1:2000, Simple Western 1:100, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen | |
| P35355 | |
| PTGS2 | |
| Recombinant protein corresponding to the C-terminus of rat COX2 (the exact amino acid sequence is considered to be proprietary information). | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Cytokine Research, Hypoxia, Lipid and Metabolism, Signal Transduction | |
| 5743 | |
| Human, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| PBS (pH 7.4), 0.2% BSA, Tween-20 with 0.05% Sodium Azide | |
| COX-2, COX2cyclooxygenase 2b, cyclooxygenase-2, EC 1.14.99, EC 1.14.99.1, GRIPGHS, hCox-2, PGG/HS, PGH synthase 2, PGHS-2, PHS II, PHS-2, prostaglandin G/H synthase 2, prostaglandin G/H synthase and cyclooxygenase, Prostaglandin H2 synthase 2, Prostaglandin-endoperoxide synthase 2, prostaglandin-endoperoxide synthase 2 (prostaglandin G/H synthase andcyclooxygenase) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This reacts with the 70 kDa protein. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction