Learn More
Bollé Safety Coverall Clear Safety Goggles
Product Code. 12697723
Change view
Click to view available options
Model:
Coverall Autoclave
Coverall Clean
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
12697723 Coverall Autoclave Each
15548281 Coverall Clean Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12697723 Supplier Bollé Safety Supplier No. COVACLAVE
  • ONLINE EXCLUSIVE PRICE
    £19.44
    £14.27 / Pack of 1
    Save £5.17
    26% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Protective goggles is specially designed to guarantee total comfort and optimum protection in controlled environments or sensitive and sterile environments such as pharmacies, micro-electronics, and operating theatres.
Coverall Autoclave:
  • High fogging resistance
  • Autoclave compatible product
  • Overflow chute for liquids
  • PLATINUM coating : anti-scratch & anti-fog
Coverall Clean:
  • Can be sterilized using gamma, beta irrdiation or etylene oxide sterilization
  • Disposable
  • Overflow chute for liquids
  • PLATINUM LITE, anti-fog coating
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Safety Goggles
Frame Color White
Model Coverall Autoclave
No. per Pack 1 Ea.
Weight (Metric) 79 g
Material Polycarbonate
Frame Material TPR