missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coronin 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£167.00 - £310.00
Specifications
| Antigen | Coronin 3 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1:50-1:100 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18195195
|
Novus Biologicals
NBP2-27289SS |
0.025 mg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18193123
|
Novus Biologicals
NBP2-27289 |
0.1 mg |
£310.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Coronin 3 Polyclonal specifically detects Coronin 3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Coronin 3 | |
| Immunohistochemistry-Paraffin 1:50-1:100 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide | |
| 23603 | |
| This antibody was developed against synthetic peptides corresponding to amino acids 407-426 of human CORO1C. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| coronin 1C, coronin, actin binding protein, 1C, coronin, actin-binding protein, 1C, coronin-1C, Coronin-3, CRNN4, HCRNN4 | |
| CORO1C | |
| IgG | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title