missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coronin 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27289SS
This item is not returnable.
View return policy
Description
Coronin 3 Polyclonal specifically detects Coronin 3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Coronin 3 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide | |
| CORO1C | |
| This antibody was developed against synthetic peptides corresponding to amino acids 407-426 of human CORO1C. | |
| 0.025 mg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Immunohistochemistry-Paraffin 1:50-1:100 | |
| coronin 1C, coronin, actin binding protein, 1C, coronin, actin-binding protein, 1C, coronin-1C, Coronin-3, CRNN4, HCRNN4 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| 23603 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction