missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PureCoat™ Carboxyl Rectangular Canted Neck Cell Culture Flasks with Vented Cap
GREENER_CHOICEBIOPHARMA

Product Code. 11907181
Click to view available options
Capacity (Metric):
250 mL
750 mL
No. per Case:
5/Cs.
50/Cs.
Unit Size:
Pack of 5
Pack of 50
This item is not returnable. View return policy

Product Code. 11907181

Brand: Corning™ 356778

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Negatively charged surface is ideal for for preparative cell culture, cell-based, and HTS screening assays

Corning™ PureCoat™ amine (positively charged) and carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 8.45 oz.
Capacity (Metric) 250 mL
Material Polystyrene
For Use With (Application) For preparative cell culture, cell-based and HTS screening assays
Neck Style Canted
No. per Case 50/Cs.
No. per Pack 5 Pk.
Shape Rectangular
Surface Coating Carboxyl
Closure Type Vented
Culture Area 75 cm2
Show More Show Less

Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.