missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butylpropane-1,3-diol, 98%, Thermo Scientific Chemicals

Product Code. 11363836
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11363836

Brand: Thermo Scientific Alfa Aesar L16927.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2819-05-8
MDL Number MFCD01075745

Specifications

Melting Point 54°C to 56°C
Boiling Point 100°C to 102°C (2 mmHg)
Molecular Formula C10H16O4
Quantity 5 g
Beilstein 1900687
Synonym ethyl 4,4-diacetylbutyrate, 4-acetyl-5-oxohexanoic acid ethyl ester, 4,4-diacetylbutyric acid ethyl ester, ethyl 4-acetyl-5-oxohexanoate, 4-acetyl-5-oxo-hexanoic acid ethyl ester, hexanoic acid,4-acetyl-5-oxo-, ethyl ester, hexanoic acid, 4-acetyl-5-oxo-, ethyl ester
InChI Key CVPCFJRJUCNUJG-VQHVLOKHSA-N
SMILES CCOC(=O)CCC(C(=O)C)C(=O)C
IUPAC Name ethyl 4-acetyl-5-oxohexanoate
Molecular Weight (g/mol) 200.23
PubChem CID 2733934
Formula Weight 132.2
Percent Purity 98%
Chemical Name or Material 2-tert-Butylpropane-1,3-diol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.