accessibility menu, dialog, popup

UserName

CH3CH3CH3HOHO

CAS RN 2819-05-8

IUPAC Name: 2-tert-butylpropane-1,3-diol
Synonyms: 2-tert-butylpropane-1,3-diol
Molecular Weight (g/mol): 132.2
Molecular Formula: C7H16O2
InChi Key: LQPVVSDKTCYYBZ-UHFFFAOYSA-N
SMILES: CC(C)(C)C(CO)CO
Boiling Point 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results