missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CELLine Disposable Bioreactor
BIOPHARMA

Product Code. 10677762 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
1000 mL
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10677762 1000 mL Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10677762 Supplier Corning™ Supplier No. 353137
  • ONLINE EXCLUSIVE PRICE
    £675.00
    £639.89 / Pack of 3
    Save £35.11
    5% Off
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Cultivation flask for secreted product, recombinant protein expression and high-yield monoclonal antibody (MAb) production

Corning™ CELLine Disposable Bioreactor for antibody and recombinant protein production.

TRUSTED_SUSTAINABILITY

Specifications

Closure Color White, Blue
Capacity (Metric) 1000 mL
For Use With (Application) Antibody and recombinant Protein production
Includes Solid White Cap for Cultivation Chamber Port, Blue Vent Cap for Nutrient Supply Chamber, Black Gasket
No. per Case 3/Cs.
No. per Pack 1/Pk.
Product Type Disposable Bioreactor
Disposable Single-use
Sterility Sterile
Closure Type Solid Cap, Vent Cap
Volume (Metric) Working 15 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.