Learn More
Corning™ BioCoat™ Tumor Invasion Multiwell Plate
Provides an environment that allows for the assessment of the invasive potential of cells in vitro
Brand: Corning™ 354167
Code : 67
Additional Details : Weight : 0.20000kg
Description
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running cell migration and invasion studies.
Specifications
0.08 cm2 | |
8 μm | |
1 | |
1/Pk. | |
Matrigel™ Matrix |
PET | |
Multiwell Plate | |
20°C | |
1/Ea. | |
96 |