missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Gelatin Rectangular Canted Neck Culture Flasks w/Vented Cap
GREENER_CHOICE

Product Code. 10125530
Change view
Click to view available options
No. per Case:
5
50
Unit Size:
Pack of 5
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Case unitSize
10125530 5 Pack of 5
10072501 50 Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10125530 Supplier Corning™ Supplier No. 354488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pre-coating with gelatin (obtained from porcine gelatin) eliminates time-consuming preparation

Corning BioCoat Gelatin cultureware provides an attachment and growth promoting substrate for the culture of a variety of cell types. Gelatin is commonly used in the culture of vascular endothelial cells, muscle, embryonic stem (ES) cells, and F9 teratocarcinoma cells. It is also suitable for promoting adhesion of transfected cell types. Gelatin is a heterogeneous mixture of water-soluble proteins derived through the hydrolysis of Collagen.

Applications include:

  • Promotion of cell attachment and spreading
  • Culture of normal and transfected F9 teratocarcinoma cells for gene expression studies
  • Culture of HUVEC for E-Selectin7 expression and VEGF induction8

Source

  • Gelatin, porcine
  • Quality Control

    • Tested for ability to promote proliferation of HUVECs
    • Tested and found negative for bacteria and fungi

    Storage

  • 4°C to 30°C under dry conditions.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Capacity (English) 8.45 oz.
    Closure Material Phenolic
    Capacity (Metric) 250 mL
    For Use With (Application) For adhesion, growth and differentiation of neuronal and transfected cell lines
    Neck Style Canted
    No. per Case 5
    Shape Rectangular
    Surface Coating Gelatin
    Sterility Sterile
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.