missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Collagen I Rectangular Canted Neck Cell Culture Flask with Vented Cap
GREENER_CHOICEBIOPHARMA

Product Code. 10441701
Change view
Click to view available options
Capacity (Metric):
70 mL
250 mL
600 mL
750 mL
Culture Area:
150 cm2
175 cm2
25 cm2
75 cm2
No. per Case:
10/Cs.
40/Cs.
5/Cs.
50/Cs.
Neck Style:
Canted
Straight
Unit Size:
Pack of 10
Pack of 40
Pack of 5
Pack of 50
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Culture Area No. per Case Neck Style unitSize
10441701 70 mL 25 cm2 10/Cs. Canted Pack of 10
10226201 600 mL 150 cm2 5/Cs. Canted Pack of 5
10175430 250 mL 75 cm2 5/Cs. Canted Pack of 5
10339370 750 mL 175 cm2 5/Cs. Straight Pack of 5
10409911 70 mL 25 cm2 50/Cs. Canted Pack of 50
10352892 600 mL 150 cm2 40/Cs. Canted Pack of 40
10133671 750 mL 175 cm2 40/Cs. Straight Pack of 40
10062501 250 mL 75 cm2 50/Cs. Canted Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 10441701 Supplier Corning™ Supplier No. 354484

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pre-coated with Collagen I for improved cell attachment and an increased proliferation rate of transfected mammalian cells, primary cells and stem cells. Corning™ BioCoat™ Collagen I Rectangular Canted Neck Cell Culture Flask with Vented Cap provides highly consistent and biologically functional precoated surfaces to more closely mimic in vivo environments for cell culture applications.

Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. in vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.

Applications include:

  • Promotion of cell attachment and spreading
  • Rapid expansion of cell populations
  • Serum-free or reduced serum culture
  • Cell adhesion assays
  • Improving survival of primary cells in culture

Source

  • Rat tail tendon
  • Quality Control

    • Tested for ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
    • Tested and found negative for bacteria and fungi
    • Collagen I purity >90% by SDS-PAGE

    Storage

  • 4°C to 30°C under dry conditions.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Barcode Yes
    Capacity (English) 2.36 oz.
    Closure Material Phenolic
    Capacity (Metric) 70 mL
    For Use With (Application) For adhesion, growth and differentiation of neuronal and transfected cell lines
    Neck Style Canted
    No. per Case 10/Cs.
    No. per Pack 10/Pk.
    Shape Rectangular
    Surface Coating Collagen I
    Sterility Sterile
    Bottom Shape Rectangular
    Closure Type Vented
    Culture Area 25 cm2
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.