missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Automation Pipette Tips for Molecular Devices FLIPR Tetra Workstation

Shop All Axygen Scientific Products


Description
Specifications
Specifications
| Volume (Metric) | 30 μL |
| Sterility | Non-sterile |
| Color | Clear |
| Format | Rack |
| Tip Style | Automation, Round |
| For Use With (Equipment) | Molecular Devices FLIPR Tetra Automation System, Hamilton MICROLAB STAR |
| Material | Polypropylene |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?