missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Automation Pipette Tips for Molecular Devices FLIPR Tetra Workstation
GREENER_CHOICEBIOPHARMA

Product Code. 15713997 Shop All Axygen Scientific Products
Click to view available options
Color:
Black
Clear
Unit Size:
Pack of 19200
This item is not returnable. View return policy

Product Code. 15713997

Brand: Axygen™ FT384BKR

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Pipette tips manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality

  • 384 Tip format
  • Polypropylene
  • Certified RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Compatibility: Molecular Devices FLIPR Tetraworkstation
  • TRUSTED_SUSTAINABILITY

    Specifications

    Volume (Metric) 30 μL
    Sterility Non-sterile
    Color Black
    Format Rack
    Tip Style Automation, Round
    For Use With (Equipment) Molecular Devices FLIPR Tetra Automation System
    Material Polypropylene
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.