missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated DYNC1H1 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
£119.00 - £295.00
Specifications
| Antigen | DYNC1H1 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235696
|
Proteintech
CL488-12345-100UL |
100 μL |
£350.00 £295.00 / 100µL Save £55.00 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237033
|
Proteintech
CL488-12345-20UL |
20 μL |
£130.00 £119.00 / 20µL Save £11.00 8% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The DYNC1H1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human DYNC1H1. This antibody recognizes human, rat, zebrafish, mouse antigen. The DYNC1H1 antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Specifications
| DYNC1H1 | |
| Immunocytochemistry/Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Human, Rat, Zebrafish, Mouse | |
| DHC1; DHC1a; DKFZp686P2245; DNCH1; DNCL; DNECL; DYHC; Dnchc1; HL-3; KIAA0325; p22 | |
| IgG | |
| Antigen affinity purification | |
| Conjugated Antibody |
| 1000 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 1778 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title