missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated DYNC1H1 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-12345-100UL
This item is not returnable.
View return policy
Description
The DYNC1H1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human DYNC1H1. This antibody recognizes human, rat, zebrafish, mouse antigen. The DYNC1H1 antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Specifications
| DYNC1H1 | |
| Polyclonal | |
| CoraLite Plus 488 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1778 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG |
| Immunocytochemistry/Immunofluorescence | |
| 1000 μg/mL | |
| DHC1; DHC1a; DKFZp686P2245; DNCH1; DNCL; DNECL; DYHC; Dnchc1; HL-3; KIAA0325; p22 | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human, Rat, Zebrafish, Mouse | |
| 532 kDa | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu