missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated Complement factor B Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
£119.00 - £295.00
Specifications
| Antigen | Complement factor B |
|---|---|
| Clone | 8D2B6 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237708
|
Proteintech
CL488-66154-20UL |
20 μL |
£130.00 £119.00 / 20µL Save £11.00 8% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234193
|
Proteintech
CL488-66154-100UL |
100 μL |
£350.00 £295.00 / 100µL Save £55.00 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Complement factor B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Complement factor B. This antibody recognizes human antigen. The Complement factor B antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.Specifications
| Complement factor B | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 629 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 8D2B6 | |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Mouse | |
| Human | |
| BF; BFD; CFAB; FB; FBI12; GBG; H2-Bf; PBF2 | |
| IgM | |
| Thiophilic affinity chromatograph | |
| Conjugated Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title