missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated Complement factor B Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66154-100UL
This item is not returnable.
View return policy
Description
The Complement factor B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Complement factor B. This antibody recognizes human antigen. The Complement factor B antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.Specifications
| Complement factor B | |
| Monoclonal | |
| 1000 μg/mL | |
| BF; BFD; CFAB; FB; FBI12; GBG; H2-Bf; PBF2 | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human | |
| 93 kDa | |
| Liquid |
| Immunofluorescence | |
| 8D2B6 | |
| CoraLite Plus 488 | |
| Mouse | |
| Thiophilic affinity chromatograph | |
| RUO | |
| 629 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction