missing translation for 'onlineSavingsMsg'
Learn More

Proteintech CoraLite™488 TUNEL Assay Apoptosis Detection Kit (green)

Product Code. p-7224555 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 Tests
50 Tests
Unit Size:
20 tests
50 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17531176 20 Tests 20 tests
17541176 50 Tests 50 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17531176 Supplier Proteintech Supplier No. PF0000620T
Add to basket
Add to basket
This item is not returnable. View return policy

A distinctive feature of apoptosis is degradation of DNA.

  • The DNA fragments tend to be vary in length in integer multiples of 180 bp-200 bp, which is shown as a ladder-like pattern in agarose gel electrophoresis.
  • This kit uses the TUNEL method, using Terminal Deoxynucleotidyl Transferase (TdT) to catalyze the incorporation of CL488-dUTP at the 3'-OH end of the broken DNA of apoptotic cells.
  • CL488-dUTP-labeled DNA can be directly observed with a fluorescence microscope or quantified with a flow cytometer.
  • The TUNEL method can selectively detect apoptotic cells, but not necrotic cells or cells with DNA strand breaks caused by irradiation and drug treatment.

Specifications

Description Emission: 515 nm
Quantity 20 Tests
Content And Storage Store at -20°C. Avoid exposure to light. Stable for 2 years after shipment. Avoid freeze- thaw cycles.
For Use With (Application) Flow Cytometry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.