missing translation for 'onlineSavingsMsg'
Learn More

Coppersensor 3, TRC

Product Code. 30132758
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132758 100 mg 100mg
30133422 50 mg 50mg
30009679 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132758 Supplier Toronto Research Chemicals Supplier No. TRCB874750100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1335151-56-8
Molecular Formula C32H56BN3O2S4
Synonym [4-Ethyl-beta-(4-ethyl-3,5-dimethyl-2H-pyrrol-2-ylidene-kN)-N,N-bis[2-[[2-(ethylthio)ethyl]thio]ethyl]-3,5-dimethyl-1H-pyrrole-2-ethanaminato-kN]dimethoxy- (T-4)- Boron∣(T-4)-[4-Ethyl-beta-(4-ethyl-3,5-dimethyl-2H-pyrrol-2-ylidene-kN)-N,N-bis[2-[[2-(ethylthio)ethyl]thio]ethyl]-3,5-dimethyl-1H-pyrrole-2-ethanaminato-kN]dimethoxyboron

Specifications

Quantity 100 mg
Shipping Condition Dry Pack
Formula Weight 653.3348
Molecular Weight (g/mol) 653.87
Chemical Name or Material Coppersensor 3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.