CAS RN 1335151-56-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Coppersensor 3, TRC
CAS: 1335151-56-8 Molecular Formula: C32H56BN3O2S4 Synonym: [4-Ethyl-beta-(4-ethyl-3,5-dimethyl-2H-pyrrol-2-ylidene-kN)-N,N-bis[2-[[2-(ethylthio)ethyl]thio]ethyl]-3,5-dimethyl-1H-pyrrole-2-ethanaminato-kN]dimethoxy- (T-4)- Boron∣(T-4)-[4-Ethyl-beta-(4-ethyl-3,5-dimethyl-2H-pyrrol-2-ylidene-kN)-N,N-bis[2-[[2-(ethylthio)ethyl]thio]ethyl]-3,5-dimethyl-1H-pyrrole-2-ethanaminato-kN]dimethoxyboron